Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACCATGGAGTCTTGTCCATCT[C/G]TCCAGTGCTGTGAAGGTTGTAGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM24A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM24A - family with sequence similarity 24 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029888.1 | 364 | Missense Mutation | CTC,GTC | L,V 79 | NP_001025059.1 | |
XM_017015638.1 | 364 | Missense Mutation | CTC,GTC | L,V 79 | XP_016871127.1 | |
XM_017015639.1 | 364 | Missense Mutation | CTC,GTC | L,V 79 | XP_016871128.1 |