Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCATAGTGGTGCTGCATGGATA[C/T]GAAGCAGTGAAGGAAGCCCTGATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601130 | ||||||||||||||||||||
Literature Links: |
CYP2C9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP2C9 - cytochrome P450 family 2 subfamily C member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000771.3 | 265 | Silent Mutation | TAC,TAT | Y,Y 80 | NP_000762.2 | |
XM_017015758.1 | 265 | Silent Mutation | TAC,TAT | Y,Y 80 | XP_016871247.1 |