Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTGAATTCAAGAAGAGTTCTAT[A/G]TATCTGTGCTGCATATTGGCCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPF - heterogeneous nuclear ribonucleoprotein F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098204.1 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_001091674.1 | |
NM_001098205.1 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_001091675.1 | |
NM_001098206.1 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_001091676.1 | |
NM_001098207.1 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_001091677.1 | |
NM_001098208.1 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_001091678.1 | |
NM_004966.3 | 1476 | Silent Mutation | TAC,TAT | Y,Y 356 | NP_004957.1 |