Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTTCATATTGAATTCCTTCAA[C/T]TCTGCGTCCTTTTAAAGGGCCCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146640 MIM: 601720 | ||||||||||||||||||||
Literature Links: |
ITIH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITIH2 - inter-alpha-trypsin inhibitor heavy chain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIN - Kin17 DNA and RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012311.3 | 1106 | Missense Mutation | ATT,GTT | I,V 381 | NP_036443.1 | |
XM_006717434.3 | 1106 | Intron | XP_006717497.1 | |||
XM_006717435.3 | 1106 | Intron | XP_006717498.1 | |||
XM_011519427.1 | 1106 | Intron | XP_011517729.1 | |||
XM_017015946.1 | 1106 | Missense Mutation | ATT,GTT | I,V 275 | XP_016871435.1 |