Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTATGTGCAGACCTACACTGCCA[G/T]GGACCTCCATGCAGACTCGCTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC283045 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC283045 - uncharacterized LOC283045 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF365 - zinc finger protein 365 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014951.2 | 1258 | Missense Mutation | AGG,ATG | R,M 132 | NP_055766.2 | |
NM_199450.2 | 1258 | Missense Mutation | AGG,ATG | R,M 132 | NP_955522.1 | |
NM_199451.2 | 1258 | Missense Mutation | AGG,ATG | R,M 132 | NP_955523.1 | |
NM_199452.3 | 1258 | Intron | NP_955524.3 | |||
XM_017015937.1 | 1258 | Missense Mutation | AGG,ATG | R,M 132 | XP_016871426.1 | |
XM_017015938.1 | 1258 | Intron | XP_016871427.1 | |||
XM_017015939.1 | 1258 | Intron | XP_016871428.1 | |||
XM_017015940.1 | 1258 | Intron | XP_016871429.1 | |||
XM_017015941.1 | 1258 | Intron | XP_016871430.1 |