Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCACCTCAGCCTGTGTGGCCGT[C/T]TATAAGGTGAGGGGCTGCGGAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611233 MIM: 610369 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDNF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDNF - cerebral dopamine neurotrophic factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029954.2 | 294 | Intron | NP_001025125.2 | |||
XM_011519488.2 | 294 | Intron | XP_011517790.1 |
HSPA14 - heat shock protein family A (Hsp70) member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278205.1 | 294 | Silent Mutation | GTC,GTT | V,V 17 | NP_001265134.1 | |
NM_016299.3 | 294 | Silent Mutation | GTC,GTT | V,V 17 | NP_057383.2 |