Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATAGCTTGGATCAGTTTAGAGC[A/G]TAGAGTTTCTTTTTCTGTATATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609248 MIM: 604479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERC4 - HECT and RLD domain containing E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278185.1 | 3482 | Missense Mutation | CGC,TGC | R,C 963 | NP_001265114.1 | |
NM_001278186.1 | 3482 | Missense Mutation | CGC,TGC | R,C 778 | NP_001265115.1 | |
NM_001278187.1 | 3482 | Intron | NP_001265116.1 | |||
NM_015601.3 | 3482 | Missense Mutation | CGC,TGC | R,C 1033 | NP_056416.2 | |
NM_022079.2 | 3482 | Missense Mutation | CGC,TGC | R,C 1041 | NP_071362.1 | |
XM_011539592.2 | 3482 | Missense Mutation | CGC,TGC | R,C 1065 | XP_011537894.1 | |
XM_011539593.2 | 3482 | Missense Mutation | CGC,TGC | R,C 1057 | XP_011537895.1 | |
XM_011539594.2 | 3482 | Missense Mutation | CGC,TGC | R,C 995 | XP_011537896.1 | |
XM_011539595.2 | 3482 | Missense Mutation | CGC,TGC | R,C 995 | XP_011537897.1 | |
XM_011539596.2 | 3482 | Missense Mutation | CGC,TGC | R,C 971 | XP_011537898.1 | |
XM_011539597.2 | 3482 | Missense Mutation | CGC,TGC | R,C 944 | XP_011537899.1 | |
XM_017016041.1 | 3482 | Missense Mutation | CGC,TGC | R,C 920 | XP_016871530.1 |
SIRT1 - sirtuin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |