Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGTAGTAGGAGGAACCTGTGGC[C/G]AGCAGGTGGTTGCCATCTGTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616467 MIM: 608071 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPCD - deleted in primary ciliary dyskinesia homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXW4 - F-box and WD repeat domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323541.1 | Intron | NP_001310470.1 | ||||
NM_022039.3 | Intron | NP_071322.1 | ||||
XM_005270053.2 | Intron | XP_005270110.2 | ||||
XM_017016570.1 | Intron | XP_016872059.1 |