Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCATTCATGATAATGTCGAACA[C/T]GGTGAGTAGATCAGTTTCTCAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BTBD16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTBD16 - BTB domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318189.1 | 278 | Missense Mutation | ACG,ATG | T,M 6 | NP_001305118.1 | |
NM_144587.3 | 278 | Missense Mutation | ACG,ATG | T,M 6 | NP_653188.2 | |
XM_011539239.2 | 278 | Missense Mutation | ACG,ATG | T,M 6 | XP_011537541.1 | |
XM_011539240.2 | 278 | Missense Mutation | ACG,ATG | T,M 6 | XP_011537542.1 | |
XM_011539241.2 | 278 | Missense Mutation | ACG,ATG | T,M 6 | XP_011537543.1 | |
XM_011539242.2 | 278 | Missense Mutation | ACG,ATG | T,M 6 | XP_011537544.1 | |
XM_017015636.1 | 278 | Missense Mutation | ACG,ATG | T,M 6 | XP_016871125.1 | |
XM_017015637.1 | 278 | UTR 5 | XP_016871126.1 |