Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACCATGTTTAGGTAGCTTGTT[A/G]TCATAATAGTGATGGCTGACAGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610139 | ||||||||||||||||||||
Literature Links: |
ST8SIA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST8SIA6 - ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004470.1 | 1075 | Silent Mutation | GAC,GAT | D,D 361 | NP_001004470.1 | |
XM_017016174.1 | 1075 | Silent Mutation | GAC,GAT | D,D 331 | XP_016871663.1 | |
XM_017016175.1 | 1075 | Intron | XP_016871664.1 |