Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTGAAGGCCATGTACCAGCCCT[C/T]GTACTTGGCATTCTGCAGCGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF8 - fibroblast growth factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206389.1 | 628 | Missense Mutation | AAG,GAG | K,E 72 | NP_001193318.1 | |
NM_006119.4 | 628 | Missense Mutation | AAG,GAG | K,E 147 | NP_006110.1 | |
NM_033163.3 | 628 | Missense Mutation | AAG,GAG | K,E 176 | NP_149353.1 | |
NM_033164.3 | 628 | Missense Mutation | AAG,GAG | K,E 165 | NP_149354.1 | |
NM_033165.3 | 628 | Missense Mutation | AAG,GAG | K,E 136 | NP_149355.1 |