Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACTACTCTTGAGGTCGGGAAGT[C/T]TGCCGGCATCCTCGAAAGCAGCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SKIDA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SKIDA1 - SKI/DACH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207371.3 | 2870 | Missense Mutation | AAA,AGA | K,R 504 | NP_997254.3 | |
XM_005252450.3 | 2870 | Missense Mutation | AAA,AGA | K,R 504 | XP_005252507.1 | |
XM_011519480.2 | 2870 | Missense Mutation | AAA,AGA | K,R 504 | XP_011517782.1 | |
XM_017016225.1 | 2870 | Missense Mutation | AAA,AGA | K,R 504 | XP_016871714.1 | |
XM_017016226.1 | 2870 | Missense Mutation | AAA,AGA | K,R 504 | XP_016871715.1 |