Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACCTGAAGAATCTGAATTTTGC[A/G]GGGAGACTGACTTCTCTAGGGGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606060 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC12 - DnaJ heat shock protein family (Hsp40) member C12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021800.2 | 422 | Missense Mutation | CCG,CTG | P,L 161 | NP_068572.1 | |
NM_201262.1 | 422 | Intron | NP_957714.1 | |||
XM_017016431.1 | 422 | Missense Mutation | CCG,CTG | P,L 79 | XP_016871920.1 | |
XM_017016432.1 | 422 | Missense Mutation | CCG,CTG | P,L 79 | XP_016871921.1 |