Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCATATTGTTTTGTTGCTTCA[A/G]TAAATCGCACAGGAATATTATATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604515 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLNK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLNK - B-cell linker | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114094.1 | 1189 | Missense Mutation | ACT,ATT | T,I 378 | NP_001107566.1 | |
NM_001258440.1 | 1189 | Intron | NP_001245369.1 | |||
NM_001258441.1 | 1189 | Intron | NP_001245370.1 | |||
NM_001258442.1 | 1189 | Intron | NP_001245371.1 | |||
NM_013314.3 | 1189 | Missense Mutation | ACT,ATT | T,I 401 | NP_037446.1 | |
XM_011539728.2 | 1189 | Missense Mutation | ACT,ATT | T,I 401 | XP_011538030.1 | |
XM_017016159.1 | 1189 | Missense Mutation | ACT,ATT | T,I 378 | XP_016871648.1 |