Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCGTCATGGACCGGTATGCAGC[C/T]GAGGTGCACAACATGGAGCAGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603845 MIM: 601926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFC2 - NADH:ubiquinone oxidoreductase subunit C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFC2-KCTD14 - NDUFC2-KCTD14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001203260.1 | 87 | Intron | NP_001190189.1 | |||
NM_001203261.1 | 87 | Intron | NP_001190190.1 | |||
NM_001203262.1 | 87 | Intron | NP_001190191.1 |
THRSP - thyroid hormone responsive | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003251.3 | 87 | Silent Mutation | GCC,GCT | A,A 22 | NP_003242.1 |