Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCAGCTGGAAAGCCACTTCCTG[A/G]GCCCGACGTAAAGGTGCCTTGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612834 MIM: 275360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHLDB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TREH - trehalase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301065.1 | 1506 | Silent Mutation | GCC,GCT | A,A 456 | NP_001287994.1 | |
NM_007180.2 | 1506 | Silent Mutation | GCC,GCT | A,A 487 | NP_009111.2 |