Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCTGCCCAGGGCAAGATGACT[A/G]TGACCGCCTGCGGCCCCTGTTCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605781 | ||||||||||||||||||||
Literature Links: |
RHOD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHOD - ras homolog family member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300886.1 | 318 | Intron | NP_001287815.1 | |||
NM_014578.3 | 318 | Missense Mutation | TAT,TGT | Y,C 78 | NP_055393.1 |