Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCTCTCCTTCACGTGGTTCTGT[A/G]CTCCCGGGCCCCCGCTGCCGGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609236 MIM: 611969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRSK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRSK2 - BR serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB2 - MOB kinase activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172223.2 | 792 | Missense Mutation | GCA,GTA | A,V 261 | NP_001165694.1 | |
NM_053005.5 | 792 | Missense Mutation | GCA,GTA | A,V 230 | NP_443731.2 |