Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGCGCCTGGGCCTGGACTTAGG[A/G]TGGAACCCTTCCGGAGAAGGCTGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602732 MIM: 616454 | ||||||||||||||||||||
Literature Links: |
ARHGAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP1 - Rho GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004308.3 | 80 | Intron | NP_004299.1 | |||
XM_011520095.1 | 80 | Intron | XP_011518397.1 | |||
XM_017017720.1 | 80 | Intron | XP_016873209.1 |
ZNF408 - zinc finger protein 408 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184751.1 | 80 | Silent Mutation | GGA,GGG | G,G 20 | NP_001171680.1 | |
NM_024741.2 | 80 | Silent Mutation | GGA,GGG | G,G 28 | NP_079017.1 |