Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134690 MIM: 606866 MIM: 608046 MIM: 603414 MIM: 604575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAU - FAU, ubiquitin like and ribosomal protein S30 fusion | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001997.4 | Intron | NP_001988.1 |
MIR6751 - microRNA 6751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL49 - mitochondrial ribosomal protein L49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004927.3 | Intron | NP_004918.1 |
SYVN1 - synoviolin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM7SF2 - transmembrane 7 superfamily member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNHIT2 - zinc finger HIT-type containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |