Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTGAAAGATACATACGTGGCG[C/T]TGAAGAGCAGAAAGCAATGAAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP4 - caspase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001225.3 | 992 | Missense Mutation | ACG,GCG | T,A 307 | NP_001216.1 | |
NM_033306.2 | 992 | Missense Mutation | ACG,GCG | T,A 251 | NP_150649.1 | |
XM_011543019.1 | 992 | Missense Mutation | ACG,GCG | T,A 216 | XP_011541321.1 | |
XM_017018397.1 | 992 | Missense Mutation | ACG,GCG | T,A 216 | XP_016873886.1 |