Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615417 MIM: 608356 MIM: 609146 MIM: 604481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BET1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BET1L - Bet1 golgi vesicular membrane trafficking protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6743 - microRNA 6743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF3 - outer dense fiber of sperm tails 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIC8A - RIC8 guanine nucleotide exchange factor A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286134.1 | 553 | Missense Mutation | ACG,ATG | T,M 12 | NP_001273063.1 | |
NM_021932.5 | 553 | Missense Mutation | ACG,ATG | T,M 12 | NP_068751.4 | |
XM_005253052.3 | 553 | Missense Mutation | ACG,ATG | T,M 12 | XP_005253109.1 | |
XM_017018107.1 | 553 | UTR 5 | XP_016873596.1 |
SIRT3 - sirtuin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |