Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGAAGTTCACGGCTCCTAGCAG[C/T]GTTTCCCCATTCTTGCATAGTAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601638 MIM: 607388 MIM: 605493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFIP2 - ADP ribosylation factor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242854.1 | 845 | Silent Mutation | ACA,ACG | T,T 231 | NP_001229783.1 | |
NM_001242855.1 | 845 | Silent Mutation | ACA,ACG | T,T 160 | NP_001229784.1 | |
NM_001242856.1 | 845 | Silent Mutation | ACA,ACG | T,T 113 | NP_001229785.1 | |
NM_012402.3 | 845 | Silent Mutation | ACA,ACG | T,T 198 | NP_036534.1 | |
XM_005252840.4 | 845 | Silent Mutation | ACA,ACG | T,T 198 | XP_005252897.1 |
TIMM10B - translocase of inner mitochondrial membrane 10 homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM3 - tripartite motif containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |