Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTGGAGGTGATGGGTAACTGG[C/G]GCCTTGGAATTGCTCCATGGGTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611642 MIM: 611641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEPACAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEPACAM - hepatic and glial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152722.4 | 514 | UTR 3 | NP_689935.2 | |||
XM_005271449.2 | 514 | UTR 3 | XP_005271506.1 | |||
XM_017017361.1 | 514 | UTR 3 | XP_016872850.1 |
HEPN1 - hepatocellular carcinoma, down-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037558.2 | 514 | Missense Mutation | CGC,GGC | R,G 5 | NP_001032647.2 |