Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTAAGCGAGAGCCTCACTGGTT[C/T]CCAGGACATCAACCTGAACTGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf1 - chromosome 11 open reading frame 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022761.2 | 475 | Silent Mutation | TTC,TTT | F,F 123 | NP_073598.1 | |
XM_011542957.2 | 475 | Silent Mutation | TTC,TTT | F,F 139 | XP_011541259.1 | |
XM_011542958.2 | 475 | Missense Mutation | TCC,TTC | S,F 117 | XP_011541260.1 | |
XM_017018129.1 | 475 | Missense Mutation | TCC,TTC | S,F 101 | XP_016873618.1 |
FDXACB1 - ferredoxin-fold anticodon binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |