Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180465 MIM: 602671 MIM: 610971 | ||||||||||||||||||||
Literature Links: |
CCDC84 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC84 - coiled-coil domain containing 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3656 - microRNA 3656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS25 - ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC37A4 - solute carrier family 37 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164277.1 | 1311 | Silent Mutation | GGC,GGT | G,G 376 | NP_001157749.1 | |
NM_001164278.1 | 1311 | Silent Mutation | GGC,GGT | G,G 398 | NP_001157750.1 | |
NM_001164279.1 | 1311 | Silent Mutation | GGC,GGT | G,G 303 | NP_001157751.1 | |
NM_001164280.1 | 1311 | Silent Mutation | GGC,GGT | G,G 376 | NP_001157752.1 | |
NM_001467.5 | 1311 | Silent Mutation | GGC,GGT | G,G 376 | NP_001458.1 |
TRAPPC4 - trafficking protein particle complex 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |