Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAGTACCTGGTGTGGCTTCAGT[A/C]TCTACCTGCTCCGGTGCCTGGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf24 - chromosome 11 open reading frame 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300913.1 | 1486 | Intron | NP_001287842.1 | |||
NM_022338.3 | 1486 | Missense Mutation | GAG,GAT | E,D 348 | NP_071733.1 | |
XM_005274053.3 | 1486 | Missense Mutation | GAG,GAT | E,D 348 | XP_005274110.1 | |
XM_011545103.2 | 1486 | Missense Mutation | GAG,GAT | E,D 316 | XP_011543405.1 |