Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCTCTTACCTTTTAAAATGTA[C/G]TCCGTTAGCAAAGTAGGCACCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FEZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FEZ1 - fasciculation and elongation protein zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005103.4 | 1429 | Silent Mutation | GAC,GAG | D,E 383 | NP_005094.1 | |
NM_022549.3 | 1429 | Intron | NP_072043.1 | |||
XM_005271734.2 | 1429 | Missense Mutation | GAC,GAG | D,E 383 | XP_005271791.1 | |
XM_005271735.2 | 1429 | Missense Mutation | GAC,GAG | D,E 383 | XP_005271792.1 |