Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCATTGTGCTGTCTCTCCAGGAC[A/G]CGTTGTCTATAATGATACAGATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602664 | ||||||||||||||||||||
Literature Links: |
CASP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASP4 - caspase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001225.3 | 1005 | Missense Mutation | GCG,GTG | A,V 311 | NP_001216.1 | |
NM_033306.2 | 1005 | Missense Mutation | GCG,GTG | A,V 255 | NP_150649.1 | |
XM_011543019.1 | 1005 | Missense Mutation | GCG,GTG | A,V 220 | XP_011541321.1 | |
XM_017018397.1 | 1005 | Missense Mutation | GCG,GTG | A,V 220 | XP_016873886.1 |