Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCGCACCAGCAGCATGGCCAG[A/G]AAGGCCACTGCCATGAGCACGCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608204 | ||||||||||||||||||||
Literature Links: |
UNC93B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UNC93B1 - unc-93 homolog B1 (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030930.3 | 583 | Silent Mutation | NP_112192.2 | |||
XM_011545290.1 | 583 | Silent Mutation | XP_011543592.1 | |||
XM_011545291.2 | 583 | Silent Mutation | XP_011543593.1 |