Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCATGCTGAGCGCCATCAGCACC[A/G]AGCGCTGCCTGTCCATCCTGTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607229 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRGPRX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRGPRX3 - MAS related GPR family member X3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_054031.3 | 571 | Intron | NP_473372.3 | |||
XM_011519882.2 | 571 | Missense Mutation | AAG,GAG | K,E 119 | XP_011518184.1 |