Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTCCAGGGAGAGCAGCAGCAGGA[A/G]GAGCCTCAGGAGATGACCCTCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612793 MIM: 173320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTDSS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTDSS2 - phosphatidylserine synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNH1 - ribonuclease/angiogenin inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002939.3 | 1532 | UTR 3 | NP_002930.2 | |||
NM_203383.1 | 1532 | UTR 3 | NP_976317.1 | |||
NM_203384.1 | 1532 | UTR 3 | NP_976318.1 | |||
NM_203385.1 | 1532 | UTR 3 | NP_976319.1 | |||
NM_203386.2 | 1532 | UTR 3 | NP_976320.1 | |||
NM_203387.2 | 1532 | UTR 3 | NP_976321.1 | |||
NM_203388.2 | 1532 | UTR 3 | NP_976322.1 | |||
NM_203389.2 | 1532 | UTR 3 | NP_976323.1 | |||
XM_011520255.1 | 1532 | UTR 3 | XP_011518557.1 | |||
XM_011520257.2 | 1532 | UTR 3 | XP_011518559.1 | |||
XM_011520258.2 | 1532 | UTR 3 | XP_011518560.1 | |||
XM_011520259.2 | 1532 | UTR 3 | XP_011518561.1 | |||
XM_011520260.2 | 1532 | UTR 3 | XP_011518562.1 | |||
XM_011520261.2 | 1532 | UTR 3 | XP_011518563.1 | |||
XM_011520262.2 | 1532 | UTR 3 | XP_011518564.1 | |||
XM_011520263.1 | 1532 | UTR 3 | XP_011518565.1 | |||
XM_017018106.1 | 1532 | UTR 3 | XP_016873595.1 |