Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCTGTGCTATCTTCTGGCTGAG[A/G]AACAAGTTCCATTCCATCACGTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C11orf65 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C11orf65 - chromosome 11 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KDELC2 - KDEL motif containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153705.4 | 1495 | Missense Mutation | CCT,TCT | P,S 486 | NP_714916.3 | |
XM_011542621.2 | 1495 | Missense Mutation | CCT,TCT | P,S 472 | XP_011540923.1 | |
XM_011542623.2 | 1495 | Missense Mutation | CCT,TCT | P,S 348 | XP_011540925.1 |