Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCTCCCCCAGCCGCCGCACCTC[C/T]TGCACCAGGCTCTGCAGAAACATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605555 MIM: 608794 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIP - aryl hydrocarbon receptor interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6752 - microRNA 6752 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM1 - phosphatidylinositol transfer protein membrane associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130848.1 | 3581 | Silent Mutation | CAA,CAG | Q,Q 1112 | NP_001124320.1 | |
NM_004910.2 | 3581 | Silent Mutation | CAA,CAG | Q,Q 1113 | NP_004901.2 | |
XM_011545396.2 | 3581 | Silent Mutation | CAA,CAG | Q,Q 1112 | XP_011543698.1 | |
XM_017018586.1 | 3581 | Silent Mutation | CAA,CAG | Q,Q 1113 | XP_016874075.1 |