Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602647 MIM: 602946 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6514 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6514 - microRNA 6514 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6748 - microRNA 6748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NXF1 - nuclear RNA export factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081491.1 | 49 | Intron | NP_001074960.1 | |||
NM_006362.4 | 49 | Intron | NP_006353.2 |
TAF6L - TATA-box binding protein associated factor 6 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM179B - transmembrane protein 179B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM223 - transmembrane protein 223 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080501.2 | 49 | Missense Mutation | ACG,ATG | T,M 10 | NP_001073970.1 |