Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTCTTTGATGATCATGGTCAAC[G/T]GTCTCATGTGCCCTGTGTTGAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615818 | ||||||||||||||||||||
Literature Links: |
DNHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNHD1 - dynein heavy chain domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144666.2 | 2270 | Missense Mutation | CGG,CTG | R,L 569 | NP_653267.2 | |
NM_173589.3 | 2270 | Missense Mutation | CGG,CTG | R,L 569 | NP_775860.3 |
RRP8 - ribosomal RNA processing 8, methyltransferase, homolog (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015324.3 | 2270 | Intron | NP_056139.1 | |||
XM_011519955.2 | 2270 | Intron | XP_011518257.1 | |||
XM_017017423.1 | 2270 | Intron | XP_016872912.1 |