Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGCCAGATCACGTCAGGTGGTG[A/G]TGAGCCCTGAGTAGTGACAGGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IGSF22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF22 - immunoglobulin superfamily member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173588.3 | 4102 | Missense Mutation | CCA,TCA | P,S 1032 | NP_775859.3 | |
XM_011520030.2 | 4102 | Missense Mutation | CCA,TCA | P,S 1052 | XP_011518332.1 | |
XM_011520031.2 | 4102 | Missense Mutation | CCA,TCA | P,S 951 | XP_011518333.1 | |
XM_017017607.1 | 4102 | Missense Mutation | CCA,TCA | P,S 1052 | XP_016873096.1 | |
XM_017017608.1 | 4102 | Missense Mutation | CCA,TCA | P,S 1032 | XP_016873097.1 |
TMEM86A - transmembrane protein 86A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |