Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCACAACAAACACGGAGCAATC[C/T]CAATGCTGTTTATCCGGAGGACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611834 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL21 - mitochondrial ribosomal protein L21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181514.1 | 634 | Missense Mutation | GAG,GGG | E,G 199 | NP_852615.1 | |
NM_181515.1 | 634 | Missense Mutation | GAG,GGG | E,G 114 | NP_852616.1 | |
XM_005273823.4 | 634 | Intron | XP_005273880.1 |