Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGATGATGCCTTACAGTGCTACTC[C/T]GAAGCTATTAAGCTGGATCCCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STIP1 - stress induced phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282652.1 | 778 | Silent Mutation | TCC,TCT | S,S 75 | NP_001269581.1 | |
NM_001282653.1 | 778 | Intron | NP_001269582.1 | |||
NM_006819.2 | 778 | Intron | NP_006810.1 |