Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGATCAAATCCCTTATAATATT[C/T]ACGGCATTTGTAAGGTTTCTCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605015 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF214 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF214 - zinc finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013249.2 | 2134 | Missense Mutation | NP_037381.2 | |||
XM_005253128.3 | 2134 | Nonsense Mutation | XP_005253185.1 | |||
XM_005253129.3 | 2134 | Nonsense Mutation | XP_005253186.1 | |||
XM_006718308.3 | 2134 | Nonsense Mutation | XP_006718371.1 | |||
XM_006718309.3 | 2134 | Nonsense Mutation | XP_006718372.1 | |||
XM_011520355.2 | 2134 | Nonsense Mutation | XP_011518657.1 | |||
XM_011520357.2 | 2134 | Nonsense Mutation | XP_011518659.1 | |||
XM_017018269.1 | 2134 | Nonsense Mutation | XP_016873758.1 |