Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTGAGCTTAGAGGAAGACCCCT[G/T]CAGGCTGGAACCAGTGTGCGAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BATF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BATF2 - basic leucine zipper ATF-like transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300807.1 | 687 | Missense Mutation | AAG,CAG | K,Q 162 | NP_001287736.1 | |
NM_001300808.1 | 687 | Missense Mutation | AAG,CAG | K,Q 101 | NP_001287737.1 | |
NM_138456.3 | 687 | Missense Mutation | AAG,CAG | K,Q 186 | NP_612465.3 |