Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGAATGGTAGGCGAGCTTATAG[C/G]CTTCTGCACCAGCACACATAAAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POLA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLA2 - DNA polymerase alpha 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002689.3 | 520 | Missense Mutation | GCC,GGC | A,G 47 | NP_002680.2 | |
XM_011544877.2 | 520 | Missense Mutation | GCC,GGC | A,G 47 | XP_011543179.1 | |
XM_011544878.2 | 520 | Missense Mutation | GCC,GGC | A,G 47 | XP_011543180.1 | |
XM_011544879.1 | 520 | Missense Mutation | GCC,GGC | A,G 47 | XP_011543181.1 | |
XM_011544881.2 | 520 | Missense Mutation | GCC,GGC | A,G 47 | XP_011543183.1 | |
XM_017017452.1 | 520 | Missense Mutation | GCC,GGC | A,G 47 | XP_016872941.1 |