Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGACAGGACCCACAGCAGGGC[A/G]CACACCACGGCCGACAGGCGCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607233 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRGPRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRGPRF - MAS related GPR family member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098515.1 | 1357 | Silent Mutation | TGC,TGT | C,C 164 | NP_001091985.1 | |
NM_145015.4 | 1357 | Silent Mutation | TGC,TGT | C,C 164 | NP_659452.3 | |
XM_017017170.1 | 1357 | Silent Mutation | TGC,TGT | C,C 164 | XP_016872659.1 |
MRGPRF-AS1 - MRGPRF antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |