Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGTGGACTTCGTCTGCCTGGAC[A/G]GTGACAGCTGCTGACTGGCCGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602635 MIM: 126452 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEAF1 - DEAF1, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293634.1 | 2348 | Silent Mutation | ACA,ACG | T,T 472 | NP_001280563.1 | |
NM_021008.3 | 2348 | Silent Mutation | ACA,ACG | T,T 547 | NP_066288.2 | |
XM_011519842.2 | 2348 | Intron | XP_011518144.1 |
DRD4 - dopamine receptor D4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |