Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCGGACAACCTCGACAGAGCCC[C/G]CCTCGGCCCTCGGGAGAGCGGCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607666 MIM: 614634 | ||||||||||||||||||||
Literature Links: |
ANGPTL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPTL5 - angiopoietin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178127.4 | 81 | Intron | NP_835228.2 | |||
XM_011542735.2 | 81 | Intron | XP_011541037.1 | |||
XM_017017466.1 | 81 | Intron | XP_016872955.1 |
CEP126 - centrosomal protein 126 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020802.3 | 81 | Missense Mutation | CCC,CGC | P,R 27 | NP_065853.3 | |
XM_005271627.4 | 81 | Missense Mutation | CCC,CGC | P,R 27 | XP_005271684.2 |