Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGAACACCACCACCGCCACGAC[A/G]CCACCGATCACGGCATGATCCACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605686 | ||||||||||||||||||||
Literature Links: |
CADM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CADM1 - cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098517.1 | 1182 | Silent Mutation | GGC,GGT | G,G 351 | NP_001091987.1 | |
NM_001301043.1 | 1182 | Silent Mutation | GGC,GGT | G,G 408 | NP_001287972.1 | |
NM_001301044.1 | 1182 | Silent Mutation | GGC,GGT | G,G 390 | NP_001287973.1 | |
NM_001301045.1 | 1182 | Silent Mutation | GGC,GGT | G,G 380 | NP_001287974.1 | |
NM_014333.3 | 1182 | Silent Mutation | GGC,GGT | G,G 379 | NP_055148.3 | |
XM_005271494.2 | 1182 | Silent Mutation | GGC,GGT | G,G 362 | XP_005271551.1 | |
XM_017017457.1 | 1182 | Silent Mutation | GGC,GGT | G,G 414 | XP_016872946.1 | |
XM_017017458.1 | 1182 | Silent Mutation | GGC,GGT | G,G 339 | XP_016872947.1 | |
XM_017017459.1 | 1182 | Silent Mutation | GGC,GGT | G,G 261 | XP_016872948.1 | |
XM_017017460.1 | 1182 | Silent Mutation | GGC,GGT | G,G 232 | XP_016872949.1 | |
XM_017017461.1 | 1182 | Silent Mutation | GGC,GGT | G,G 204 | XP_016872950.1 |