Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTCCAAGGCCCTGGAGCTGCAG[C/G]GGGTGATGGCAGCAGAGGCTGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159555 MIM: 613934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT2A - lysine methyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929089 - uncharacterized LOC101929089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM25 - transmembrane protein 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC36 - tetratricopeptide repeat domain 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080441.1 | 172 | Missense Mutation | CGG,GGG | R,G 58 | NP_001073910.1 |