Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTATGGCCCAAATGTCATTGAC[C/T]ATTTCTCCTGTGATGCCTCACCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR6Q1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR6Q1 - olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005186.2 | 538 | Missense Mutation | CAT,TAT | H,Y 180 | NP_001005186.2 |
OR9Q1 - olfactory receptor family 9 subfamily Q member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005212.3 | 538 | Intron | NP_001005212.1 |
VN2R9P - vomeronasal 2 receptor 9 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |