Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCCCTGACTCTCAAGTTGCTG[C/T]CCACAGCCACTGCCTCTTGCTCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612880 | ||||||||||||||||||||
Literature Links: |
CCDC89 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC89 - coiled-coil domain containing 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152723.2 | 1112 | Missense Mutation | GAC,GGC | D,G 322 | NP_689936.1 |
SYTL2 - synaptotagmin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |