Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGCCTTCGACCGCACAGAGACC[C/T]GCACAGCTTACCCAAATGCCTTTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNGA4 - cyclic nucleotide gated channel alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037329.3 | 500 | Missense Mutation | CGC,TGC | R,C 160 | NP_001032406.1 | |
XM_017017217.1 | 500 | Missense Mutation | CGC,TGC | R,C 160 | XP_016872706.1 | |
XM_017017218.1 | 500 | Missense Mutation | CGC,TGC | R,C 120 | XP_016872707.1 | |
XM_017017219.1 | 500 | Missense Mutation | CGC,TGC | R,C 120 | XP_016872708.1 |
FAM160A2 - family with sequence similarity 160 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |